4-Chlorthioanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
C1156-10G
C1156-25G
TCIAC1156-10GEA
321
DKK
TCIAC1156-10G
TCIAC1156-25G
4-Chlorthioanisol ≥98.0% (by GC)
4-Chlorthioanisol
Formel:
C₇H₇ClS Molekylvægt: 158.65 g/mol Kogepunkt: 224°C |
MDL Number:
MFCD00013643 CAS-nummer: 123-09-1 EINECS: 204-600-5 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...