3-Chlor-5-Fluoranisol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-3-fluoro-5-methoxybenzene
C2218-1G
C2218-5G
TCIAC2218-1GEA
288
DKK
TCIAC2218-1G
TCIAC2218-5G
3-Chlor-5-Fluoranisol ≥96.0% (by GC)
3-Chlor-5-Fluoranisol
Formel:
C₇H₆ClFO Flammepunkt: 79°C |
MDL Number:
MFCD00070770 CAS-nummer: 202925-08-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...