3'-Chloracetophenon ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
m-Chloroacetophenone, 1-(3-Chlorophenyl)ethanone
C0096-10ML
C0096-25ML
TCIAC0096-10MLEA
411
DKK
TCIAC0096-10ML
TCIAC0096-25ML
3'-Chloracetophenon ≥97.0% (by GC)
3'-Chloracetophenon
Formel:
C₈H₇ClO Molekylvægt: 154.6 g/mol Kogepunkt: 230°C Flammepunkt: 105°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000593 CAS-nummer: 99-02-5 EINECS: 202-721-8 UN nummer: 3416 ADR: 6.1 II |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...