4-Chlor-m-xylen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2165-25G
C2165-5G
TCIAC2165-25GEA
1020
DKK
TCIAC2165-25G
TCIAC2165-5G
4-Chlor-m-xylen ≥98.0% (by GC)
4-Chlor-m-xylen
Formel:
C₈H₉Cl Kogepunkt: 188°C Smeltepunkt: –32°C |
MDL Number:
MFCD00060644 CAS-nummer: 95-66-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...