4-Brom-N,N-dimethylanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0585-25G B0585-500G B0585-100G
TCIAB0585-25GEA 173 DKK
TCIAB0585-25G TCIAB0585-500G TCIAB0585-100G
4-Brom-N,N-dimethylanilin ≥98.0% (by GC)
4-Brom-N,N-dimethylanilin
Formel: C₈H₁₀BrN
Kogepunkt: 264°C
Smeltepunkt: 54…55 °C
Massefylde: 1,3218 g/cm³ (99.5 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000093
CAS-nummer: 586-77-6
EINECS: 209-582-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1e8c603d5e9012be67435c46712a6ac8040935c&vid=e1e8c603d5e9012be67435c46712a6ac8040935c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR