4-Brom-α,α,α,2-tetrafluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromo-2-fluorobenzotrifluoride, 1-Bromo-3-fluoro-4-(trifluoromethyl)benzene

B2946-5G B2946-25G
TCIAB2946-5GEA 238 DKK
TCIAB2946-5G TCIAB2946-25G
4-Brom-α,α,α,2-tetrafluortoluen ≥98.0% (by GC)
4-Brom-α,α,α,2-tetrafluortoluen
Formel: C₇H₃BrF₄
MDL Number: MFCD01075254
CAS-nummer: 142808-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d63fd10449ab2916122249d9bc91027c0ae7a978&vid=d63fd10449ab2916122249d9bc91027c0ae7a978&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR