3'-Bromacetophenon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
m-Bromoacetophenone
B0536-25G
B0536-5G
TCIAB0536-25GEA
288
DKK
TCIAB0536-25G
TCIAB0536-5G
3'-Bromacetophenon ≥98.0% (by GC)
3'-Bromacetophenon
Formel:
C₈H₇BrO Kogepunkt: 136°C Massefylde: 1,49 g/cm³ (20 °C) Flammepunkt: 114°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000083 CAS-nummer: 2142-63-4 EINECS: 218-396-0 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...