p-Toluidin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Toluidin

T0300-25G T0300-500G
TCIAT0300-25GEA 132 DKK
TCIAT0300-25G TCIAT0300-500G
p-Toluidin ≥99.0% (by GC, titration analysis)
p-Toluidin
Formel: C₇H₉N
Molekylvægt: 107.16 g/mol
Kogepunkt: 201
Smeltepunkt: 43
Flammepunkt: 87
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007906
CAS-nummer: 106-49-0
EINECS: 203-403-1
UN nummer: 3451
ADR: 6.1 II
Merck Index: 14,09536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ce28aa2cfa53138b50e0a081f05fa5a9fdd79d8&vid=9ce28aa2cfa53138b50e0a081f05fa5a9fdd79d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in dilute HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR