4-Fluor-o-toluidin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F0398-25G
F0398-5G
TCIAF0398-25GEA
567
DKK
TCIAF0398-25G
TCIAF0398-5G
4-Fluor-o-toluidin ≥98.0% (by GC, titration analysis)
4-Fluor-o-toluidin
Formel:
C₇H₈FN Kogepunkt: 102°C Smeltepunkt: 14 °C Massefylde: 1,1263 g/cm³ (18 °C) Flammepunkt: 88°C |
MDL Number:
MFCD00007832 CAS-nummer: 452-71-1 EINECS: 207-208-2 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...