4'-Aminoacetophenon ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A0251-25G
A0251-100G
TCIAA0251-25GEA
370
DKK
TCIAA0251-25G
TCIAA0251-100G
4'-Aminoacetophenon ≥98.0% (by titrimetric analysis)
4'-Aminoacetophenon
Formel:
C₈H₉NO Kogepunkt: 293 °C (1013 hPa) Smeltepunkt: 101…103 °C Massefylde: 1,13 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007896 CAS-nummer: 99-92-3 EINECS: 202-801-2 Merck Index: 14,00413 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 105.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...