2-Amino-5-Chlorphenol 97,0 GC, T

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Chloro-2-hydroxyaniline

A1958-25G A1958-5G
TCIAA1958-25GEA 550 DKK
TCIAA1958-25G TCIAA1958-5G
2-Amino-5-Chlorphenol 97,0 GC, T
2-Amino-5-Chlorphenol
Formel: C₆H₆ClNO
Molekylvægt: 143.57 g/mol
Smeltepunkt: 155°C
MDL Number: MFCD02093863
CAS-nummer: 28443-50-7
EINECS: 249-020-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7f0b360046dad16ddd95f3f5330b4280e5fc966&vid=a7f0b360046dad16ddd95f3f5330b4280e5fc966&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystalline
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR