2-Amino-4-fluorphenol ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Fluoro-2-hydroxyaniline

A2400-25G A2400-5G
TCIAA2400-25GEA 9050 DKK
TCIAA2400-25G TCIAA2400-5G
2-Amino-4-fluorphenol ≥97.0% (by titrimetric analysis)
2-Amino-4-fluorphenol
Formel: C₆H₆FNO
Smeltepunkt: 146°C
MDL Number: MFCD00077451
CAS-nummer: 399-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e9cd73ea147761c467656cf59f4694ff1e89d29&vid=3e9cd73ea147761c467656cf59f4694ff1e89d29&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR