3-Chlor-p-toluidin ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Chloro-p-methylaniline, 2-Chloro-4-aminotoluene, 4-Amino-2-chlorotoluene, 3-Chloro-4-methylaniline

C0538-25G C0538-500G
TCIAC0538-25GEA 304 DKK
TCIAC0538-25G TCIAC0538-500G
3-Chlor-p-toluidin ≥97.0% (by GC)
3-Chlor-p-toluidin
Formel: C₇H₈ClN
Molekylvægt: 141.6 g/mol
Kogepunkt: 244°C
Smeltepunkt: 26
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007773
CAS-nummer: 95-74-9
EINECS: 202-446-3
UN nummer: 2239
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1891b0c4c85f572d70ebc21a287bffb82a8c0df8&vid=1891b0c4c85f572d70ebc21a287bffb82a8c0df8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR