3-Brom-o-toluidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

B3482-25G B3482-5G
TCIAB3482-25GEA 3020 DKK
TCIAB3482-25G TCIAB3482-5G
3-Brom-o-toluidin ≥98.0% (by GC, titration analysis)
3-Brom-o-toluidin
Formel: C₇H₈BrN
Kogepunkt: 255°C
Massefylde: 1,51 g/cm³ (20 °C)
MDL Number: MFCD00051579
CAS-nummer: 55289-36-6
EINECS: 259-568-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c53ffb8292c3fb3af82302c05c1b2b1d1108c29b&vid=c53ffb8292c3fb3af82302c05c1b2b1d1108c29b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR