4,5-Difluor-2-nitroanilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,4-Difluoro-6-nitroaniline
D3644-25G
D3644-5G
TCIAD3644-25GEA
1480
DKK
TCIAD3644-25G
TCIAD3644-5G
4,5-Difluor-2-nitroanilin ≥98.0% (by GC)
4,5-Difluor-2-nitroanilin
Formel:
C₆H₄F₂N₂O₂ Molekylvægt: 174.11 g/mol |
MDL Number:
MFCD00010876 CAS-nummer: 78056-39-0 |
|
Specifikationer Testresultater
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...