2-Chlor-6-(trifluormethyl)anilin ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-3-chloro-α,α,α-trifluortoluene, 2-Amino-3-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-o-toluidine, 2-Amino-3-chlorobenzotrifluoride, 2-Chloro-6-trifluoromethylaniline

A1510-25G A1510-5G
TCIAA1510-25GEA 821 DKK
TCIAA1510-25G TCIAA1510-5G
2-Chlor-6-(trifluormethyl)anilin ≥97.0% (by GC)
2-Chlor-6-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Molekylvægt: 195.57 g/mol
Kogepunkt: 40°C
Flammepunkt: 71°C
MDL Number: MFCD00272565
CAS-nummer: 433-94-3
EINECS: 207-091-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f03f97f1b24dd68d42c81b27e728e63d9f7b557&vid=1f03f97f1b24dd68d42c81b27e728e63d9f7b557&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR