2-Chlor-5-(trifluormethyl)anilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Amino-4-chloro-α,α,α-trifluortoluene, 3-Amino-4-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-4-chlorobenzotrifluoride

A0948-25G A0948-500G
TCIAA0948-25GEA 279 DKK
TCIAA0948-25G TCIAA0948-500G
2-Chlor-5-(trifluormethyl)anilin ≥98.0% (by GC)
2-Chlor-5-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Kogepunkt: 106°C
Smeltepunkt: 10 °C
Massefylde: 1,42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007673
CAS-nummer: 121-50-6
EINECS: 204-475-7
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fafab5547588aa16e281f270c26c781c5400ee3&vid=2fafab5547588aa16e281f270c26c781c5400ee3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR