2-Chlormethyl-m-xylen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: α-Chloro-1,2,6-trimethylbenzene, 1-(Chloromethyl)-2,6-dimethylbenzene, 2,6-Dimethylbenzyl chloride, 2-(Chloromethyl)-1,3-dimethylbenzene

D3501-25G D3501-5G
TCIAD3501-25GEA 1670 DKK
TCIAD3501-25G TCIAD3501-5G
2-Chlormethyl-m-xylen ≥98.0% (by GC)
2-Chlormethyl-m-xylen
Formel: C₉H₁₁Cl
Kogepunkt: 100°C
MDL Number: MFCD01310815
CAS-nummer: 5402-60-8
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e74a9da1a5b00bd61a63d1b28a322c15e25aa&vid=8a1e74a9da1a5b00bd61a63d1b28a322c15e25aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR