Dibutylamin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Di-n-butylamine, N-butyl-1-propanamine

D0218-25ML D0218-500ML
TCIAD0218-25MLEA 132 DKK
TCIAD0218-25ML TCIAD0218-500ML
Dibutylamin ≥99.0% (by GC, titration analysis)
Dibutylamin
Formel: C₈H₁₉N
Kogepunkt: 159°C
Smeltepunkt: -59°C
Massefylde: 0,759 g/cm³ (20 °C)
Flammepunkt: 43°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009429
CAS-nummer: 111-92-2
EINECS: 203-921-8
UN nummer: 2248
ADR: 8,II
Merck Index: 14,03032
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab1184f23fb3f770d58f45690f3311c0bcfebab&vid=4ab1184f23fb3f770d58f45690f3311c0bcfebab&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR