1-sec-Butyl-4-nitrobenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(sec-Butyl)-4-nitrobenzene

B2428-25G B2428-5G
TCIAB2428-25GEA 2070 DKK
TCIAB2428-25G TCIAB2428-5G
1-sec-Butyl-4-nitrobenzen ≥97.0% (by GC)
1-sec-Butyl-4-nitrobenzen
Formel: C₁₀H₁₃NO₂
Kogepunkt: 144°C
MDL Number: MFCD00130025
CAS-nummer: 4237-40-5
EINECS: 224-196-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa071b4b18ee37d83b98c21cf6b11c109ac87c7&vid=afa071b4b18ee37d83b98c21cf6b11c109ac87c7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR