1,7-Octadien ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Octa-1,7-diene

O0104-25ML O0104-500ML O0104-100ML
TCIAO0104-25MLEA 197 DKK
TCIAO0104-25ML TCIAO0104-500ML TCIAO0104-100ML
1,7-Octadien ≥97.0% (by GC)
1,7-Octadiene
Formel: C₈H₁₄
Kogepunkt: 120°C
Smeltepunkt: -70°C
Massefylde: 0,7313 g/cm³ (20 °C)
Flammepunkt: 9°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008668
CAS-nummer: 3710-30-3
EINECS: 223-054-9
UN nummer: 2309
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf60328f2f0de48026396298952df0fdaa783379&vid=cf60328f2f0de48026396298952df0fdaa783379&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR