1,4-benzendithiol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzene-1,4-dithiol

B1505-1G B1505-5G
TCIAB1505-1GEA 952 DKK
TCIAB1505-1G TCIAB1505-5G
1,4-benzendithiol ≥98.0% (by GC)
1,4-benzendithiol
Formel: C₆H₆S₂
Kogepunkt: 240 °C (1013 hPa)
Smeltepunkt: 100…101 °C
MDL Number: MFCD00142541
CAS-nummer: 624-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10d04a79fbf80d677abf60b3c8039cbea5a2f7a&vid=a10d04a79fbf80d677abf60b3c8039cbea5a2f7a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 95.0 %
Melting point 96.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR