Dimethyl terephthalat ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Terephthalic acid dimethyl ester
T0015-25G
T0015-500G
TCIAT0015-25GEA
156
DKK
TCIAT0015-25G
TCIAT0015-500G
Dimethyl terephthalat ≥99.0% (by GC)
Dimethyl terephthalat
Formel:
C₁₀H₁₀O₄ Kogepunkt: 288 °C (1013 hPa) Smeltepunkt: 139 °C Massefylde: 1,2 g/cm³ (20 °C) Flammepunkt: 141 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008440 CAS-nummer: 120-61-6 EINECS: 204-411-8 Merck Index: 12,09306 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 141.0 to 144.0 deg-C |
Solubility in hot Methanol | almost transparency |
Acid Value | max. 0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...