3',4'-Difluoracetophenon ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone
D1906-25G
D1906-5G
TCIAD1906-25GEA
838
DKK
TCIAD1906-25G
TCIAD1906-5G
3',4'-Difluoracetophenon ≥97.0% (by GC)
3',4'-Difluoracetophenon
Formel:
C₈H₆F₂O Molekylvægt: 156.13 g/mol Kogepunkt: 94°C Smeltepunkt: 20 Massefylde: 1.24 Flammepunkt: 75°C |
MDL Number:
MFCD00009891 CAS-nummer: 369-33-5 EINECS: 206-717-7 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...