Diethylsebacat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate
S0024-25ML
S0024-500ML
TCIAS0024-25MLEA
132
DKK
TCIAS0024-25ML
TCIAS0024-500ML
Diethylsebacat ≥98.0% (by GC)
Diethylsebacat
Formel:
C₁₄H₂₆O₄ Kogepunkt: 309°C Smeltepunkt: –3 °C Massefylde: 0,9646 g/cm³ (20 °C) Flammepunkt: 167°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009219 CAS-nummer: 110-40-7 EINECS: 203-764-5 Merck Index: 14,08415 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...