(S)-(+)-1-Aminopropan-2-ol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0975-1G
A0975-5G
TCIAA0975-1GEA
255
DKK
TCIAA0975-1G
TCIAA0975-5G
(S)-(+)-1-Aminopropan-2-ol ≥98.0% (by GC, titration analysis)
(S)-(+)-1-Aminopropan-2-ol
Formel:
C₃H₉NO Kogepunkt: 160°C Smeltepunkt: –2 °C Massefylde: 0,96 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064429 CAS-nummer: 2799-17-9 EINECS: 220-533-4 UN nummer: 3263 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 99.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...