(R)-(-)-α-Methoxyphenyleddikesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: (R)-(-)-alpha-Methoxyphenylacetic Acid
M0830-1G M0830-100MG M0830-5G
TCIAM0830-1GEA 362 DKK
TCIAM0830-1G TCIAM0830-100MG TCIAM0830-5G
(R)-(-)-α-Methoxyphenyleddikesyre ≥98.0% (by GC, titration analysis)
(R)-(-)-α-Methoxyphenyleddikesyre
Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
Kogepunkt: 130°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004250
CAS-nummer: 3966-32-3
EINECS: 223-580-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e61408107d45026a9fa2e9fc889ee01365237e5&vid=1e61408107d45026a9fa2e9fc889ee01365237e5&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 70.0 deg-C