(R)-(+)-3-Butyn-2-ol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne

B2909-1G B2909-5G
TCIAB2909-1GEA 805 DKK
TCIAB2909-1G TCIAB2909-5G
(R)-(+)-3-Butyn-2-ol ≥98.0% (by GC)
(R)-(+)-3-Butyn-2-ol
Formel: C₄H₆O
Kogepunkt: 103°C
Massefylde: 0,891 g/cm³ (20 °C)
Flammepunkt: 26°C
MDL Number: MFCD00211237
CAS-nummer: 42969-65-3
UN nummer: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41d3132b98ee0a3fa8bcd5f733f4d7a4893bdf26&vid=41d3132b98ee0a3fa8bcd5f733f4d7a4893bdf26&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D +44.0 to +49.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR