(4-Hydroxyphenyl)eddikesyre ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-Hydroxyphenylacetic Acid
H0290-25G H0290-500G H0290-100G
TCIAH0290-25GEA 132 DKK
TCIAH0290-25G TCIAH0290-500G TCIAH0290-100G
(4-Hydroxyphenyl)eddikesyre ≥99.0% (by GC, titration analysis)
(4-Hydroxyphenyl)eddikesyre
Formel: (C₆H₄(OH))CH₂COOH
Molekylvægt: 152.15 g/mol
Kogepunkt: 347
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004347
CAS-nummer: 156-38-7
EINECS: 205-851-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff306a7d22d97fbae3f35eea6bca0c376dfa515e&vid=ff306a7d22d97fbae3f35eea6bca0c376dfa515e&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 149.0 to 153.0 deg-C
Solubility in Methanol almost transparency