(3-fluorphenyl)eddikesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(3-Fluorophenyl)acetic acid

F0216-25G F0216-5G
TCIAF0216-25GEA 928 DKK
TCIAF0216-25G TCIAF0216-5G
(3-fluorphenyl)eddikesyre ≥98.0% (by GC, titration analysis)
(3-fluorphenyl)eddikesyre
Formel: (C₆H₄F)CH₂COOH
Kogepunkt: 125°C
MDL Number: MFCD00004331
CAS-nummer: 331-25-9
EINECS: 206-360-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d467a9efafc47a89531d16fc32394b4fc24134a8&vid=d467a9efafc47a89531d16fc32394b4fc24134a8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR