(3,4-Dichlorphenyl)eddikesyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,4-Dichlorophenylacetic acid
D1813-25G
D1813-500G
TCIAD1813-25GEA
296
DKK
TCIAD1813-25G
TCIAD1813-500G
(3,4-Dichlorphenyl)eddikesyre ≥98.0% (by HPLC, titration analysis)
(3,4-Dichlorphenyl)eddikesyre
Formel:
(C₆H₃Cl₂)CH₂COOH Kogepunkt: 314 °C (1013 hPa) Smeltepunkt: 91…92 °C |
MDL Number:
MFCD00004333 CAS-nummer: 5807-30-7 EINECS: 227-368-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 88.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...