(2-Chlorphenyl)edikkesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0916-25G C0916-500G
TCIAC0916-25GEA 132 DKK
TCIAC0916-25G TCIAC0916-500G
(2-Chlorphenyl)edikkesyre ≥98.0% (by titrimetric analysis)
(2-Chlorphenyl)edikkesyre
Formel: (C₆H₄Cl)CH₂COOH
Smeltepunkt: 93…96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004317
CAS-nummer: 2444-36-2
EINECS: 219-482-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b2201effeb7bf5f14ae7ea46b8ed2e64684ed57&vid=3b2201effeb7bf5f14ae7ea46b8ed2e64684ed57&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 94.0 to 98.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR