(2-Chlor-6-fluorphenyl)eddikesyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C1126-25G
C1126-5G
TCIAC1126-25GEA
714
DKK
TCIAC1126-25G
TCIAC1126-5G
(2-Chlor-6-fluorphenyl)eddikesyre ≥98.0% (by titrimetric analysis)
(2-Chlor-6-fluorphenyl)eddikesyre
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 120.0 to 125.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...