2-Methylindol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0346-25G M0346-100G
TCIAM0346-25GEA 288 DKK
TCIAM0346-25G TCIAM0346-100G
2-Methylindol ≥99.0% (by GC)
2-Methylindole
Formel: C₉H₉N
Kogepunkt: 272°C
Smeltepunkt: 56…59 °C
Massefylde: 1,07 g/cm³ (20 °C)
Flammepunkt: 141 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005616
CAS-nummer: 95-20-5
EINECS: 202-398-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dea062ac84aeb1e9a50186b4afa1e81d9efb2ec&vid=8dea062ac84aeb1e9a50186b4afa1e81d9efb2ec&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 58.0 to 61.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR