1,3-Benzodioxol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene

M0658-25ML M0658-500ML M0658-100ML
TCIAM0658-25MLEA 197 DKK
TCIAM0658-25ML TCIAM0658-500ML TCIAM0658-100ML
1,3-Benzodioxol ≥99.0% (by GC)
1,3-Benzodioxol
Formel: C₇H₆O₂
Kogepunkt: 172°C
Smeltepunkt: –18 °C
Massefylde: 1,18 g/cm³ (20 °C)
Flammepunkt: 60°C
MDL Number: MFCD00005818
CAS-nummer: 274-09-9
EINECS: 205-992-0
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbfbe0c04c76d5ee1da39db5abeb9fa88ce508f6&vid=bbfbe0c04c76d5ee1da39db5abeb9fa88ce508f6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR