Diethylsulfat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D0525-500ML D0525-25ML
TCIAD0525-500MLEA 230 DKK
TCIAD0525-500ML TCIAD0525-25ML
Diethylsulfat ≥98.0% (by GC)
Diethylsulfat
Formel: C₄H₁₀O₄S
Kogepunkt: 208°C
Smeltepunkt: -25°C
Massefylde: 1,179 g/cm³ (20 °C)
Flammepunkt: 110°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009099
CAS-nummer: 64-67-5
EINECS: 200-589-6
UN nummer: 1594
ADR: 6.1,II
Merck Index: 14,03130
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad2bd429f6edbd5958c5b244be4c8c9a52543149&vid=ad2bd429f6edbd5958c5b244be4c8c9a52543149&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR