3-Methyl-3-buten-1-ol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M0726-25ML
M0726-500ML
M0726-100ML
TCIAM0726-25MLEA
132
DKK
TCIAM0726-25ML
TCIAM0726-500ML
TCIAM0726-100ML
3-Methyl-3-buten-1-ol ≥98.0% (by GC)
3-Methyl-3-buten-1-ol
Formel:
C₅H₁₀O Kogepunkt: 130°C Smeltepunkt: –82 °C Massefylde: 0,853 g/cm³ (20 °C) Flammepunkt: 42°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002933 CAS-nummer: 763-32-6 EINECS: 212-110-8 UN nummer: 1987 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...