Trimethoxyvinylsilan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Trimethoxy(vinyl)silane

V0042-25ML V0042-500ML V0042-100ML
TCIAV0042-25MLEA 132 DKK
TCIAV0042-25ML TCIAV0042-500ML TCIAV0042-100ML
Trimethoxyvinylsilan ≥98.0% (by GC)
Trimethoxyvinylsilan
Formel: C₅H₁₂O₃Si
Kogepunkt: 123°C
Smeltepunkt: 0 °C
Massefylde: 0,97 g/cm³ (20 °C)
Flammepunkt: 26°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008605
CAS-nummer: 2768-02-7
EINECS: 220-449-8
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74518cb5aedbb4c0f6335d0cd06bb30cb832704a&vid=74518cb5aedbb4c0f6335d0cd06bb30cb832704a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR