Tributyl borat ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tri-n-butylborate

B0518-25ML B0518-500ML B0518-100ML
TCIAB0518-25MLEA 132 DKK
TCIAB0518-25ML TCIAB0518-500ML TCIAB0518-100ML
Tributyl borat ≥99.0% (by titrimetric analysis)
Tributyl borat
Formel: C₁₂H₂₇BO₃
Kogepunkt: 230°C
Smeltepunkt: –70 °C
Massefylde: 0,8567 g/cm³ (20 °C)
Flammepunkt: 93°C
MDL Number: MFCD00009434
CAS-nummer: 688-74-4
EINECS: 211-706-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29363d25e58460361a5e431f3ea03cc5c6fabc4&vid=d29363d25e58460361a5e431f3ea03cc5c6fabc4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR