N-Ethyl-m-toluidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-Ethyl-3-methylaniline, 3-(Ethylamino)toluene

E0178-25ML E0178-500ML
TCIAE0178-25MLEA 132 DKK
TCIAE0178-25ML TCIAE0178-500ML
N-Ethyl-m-toluidin ≥98.0% (by GC, titration analysis)
N-Ethyl-m-toluidin
Formel: C₉H₁₃N
Molekylvægt: 135.21 g/mol
Kogepunkt: 221°C
Smeltepunkt: 9
Flammepunkt: 104°C
MDL Number: MFCD00009027
CAS-nummer: 102-27-2
EINECS: 203-019-4
UN nummer: 2754
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57a0f49ebbea7b80728160debe816c03504b2996&vid=57a0f49ebbea7b80728160debe816c03504b2996&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR