3-Iodbenzoylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0720-1G I0720-5G
TCIAI0720-1GEA 362 DKK
TCIAI0720-1G TCIAI0720-5G
3-Iodbenzoylchlorid ≥98.0% (by GC, titration analysis)
3-Iodbenzoylchlorid
Formel: C₇H₄ClIO
Kogepunkt: 146°C
Smeltepunkt: 23…25 °C
MDL Number: MFCD02093934
CAS-nummer: 1711-10-0
EINECS: 216-979-4
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90bb498f8d0f817c3653eecd63970b6d8971dda4&vid=90bb498f8d0f817c3653eecd63970b6d8971dda4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR