3-(Trifluormethyl)kanelsyre ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 3-(3-(Trifluoromethyl)phenyl)acrylic acid
T1547-25G T1547-5G
TCIAT1547-25GEA 2610 DKK
TCIAT1547-25G TCIAT1547-5G
3-(Trifluormethyl)kanelsyre ≥99.0% (by GC, titration analysis)
3-(Trifluormethyl)kanelsyre
Formel: C₁₀H₇F₃O₂
Molekylvægt: 216.16 g/mol
Kogepunkt: 292
MDL Number: MFCD00004393
CAS-nummer: 779-89-5
EINECS: 212-301-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46b5361e380dc090cbc24f3c4332ff8aa33bee12&vid=46b5361e380dc090cbc24f3c4332ff8aa33bee12&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 133.0 to 136.0 deg-C
Solubility in Methanol very faint turbidity