2,3,6-Trifluorbenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
T1456-25G T1456-5G
TCIAT1456-25GEA 2920 DKK
TCIAT1456-25G TCIAT1456-5G
2,3,6-Trifluorbenzoesyre ≥98.0% (by GC, titration analysis)
2,3,6-Trifluorbenzoesyre
Formel: C₇H₃F₃O₂
Molekylvægt: 176.09 g/mol
Kogepunkt: 237
MDL Number: MFCD00002409
CAS-nummer: 2358-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb073a170d4da1f7124653c92fd50282a922c95&vid=afb073a170d4da1f7124653c92fd50282a922c95&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 131.0 deg-C
Solubility in Methanol very faint turbidity