D-(-)-Glutamic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

G0057-25G G0057-250G
TCIAG0057-25GEA 189 DKK
TCIAG0057-25G TCIAG0057-250G
D-(-)-Glutamic acid ≥98.0% (by titrimetric analysis)
D-(-)-Glutamic acid
Formel: C₅H₉NO₄
Kogepunkt: 325 °C (1013 hPa)
Smeltepunkt: 213 °C
Massefylde: 1,533 g/cm³ (20 °C)
MDL Number: MFCD00063112
CAS-nummer: 6893-26-1
EINECS: 230-000-8
Merck Index: 14,04469
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb48bb7a6c8637bd30e8cd976bc918253026dbd4&vid=fb48bb7a6c8637bd30e8cd976bc918253026dbd4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -30.0 to -33.0 deg (C=5, HCl (1+5))
Solubility in HCl (1+10) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR