Calcium D-gluconat monohydrat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate
G0037-25G
G0037-500G
TCIAG0037-25GEA
132
DKK
TCIAG0037-25G
TCIAG0037-500G
Calcium D-gluconat monohydrat ≥98.0% (by titrimetric analysis)
Calcium D-gluconat monohydrat
Formel:
C₁₂H₂₂CaO₁₄·H₂O Molekylvægt: 448.39 g/mol Smeltepunkt: 178 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00150787 CAS-nummer: 299-28-5 EINECS: 206-075-8 Merck Index: 14,01669 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...