(3-Hydroxyphenyl)eddikesyre ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Hydroxyphenylacetic acid

H0356-25G H0356-5G
TCIAH0356-25GEA 1210 DKK
TCIAH0356-25G TCIAH0356-5G
(3-Hydroxyphenyl)eddikesyre ≥98.0% (by HPLC, titration analysis)
(3-Hydroxyphenyl)eddikesyre
Formel: (C₆H₄(OH))CH₂COOH
Kogepunkt: 349 °C (1013 hPa)
Smeltepunkt: 129…133 °C
Massefylde: 1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004337
CAS-nummer: 621-37-4
EINECS: 210-684-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7921d9d64c1cdaeca300ff8391f10b7376dc8077&vid=7921d9d64c1cdaeca300ff8391f10b7376dc8077&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR