2,2-Dimethylcapronsyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,2-Dimethylhexanoic acid
D2809-1G
D2809-5G
TCIAD2809-1GEA
920
DKK
TCIAD2809-1G
TCIAD2809-5G
2,2-Dimethylcapronsyre ≥98.0% (by GC, titration analysis)
2,2-Dimethylcapronsyre
Formel:
C₈H₁₆O₂ Molekylvægt: 144.21 g/mol Kogepunkt: 122°C |
MDL Number:
MFCD00965097 CAS-nummer: 813-72-9 UN nummer: 3265 ADR: 8 III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...