Methylsalicylat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Wintergreen Oil, Methyl 2-hydroxybenzoate

S0015-25G S0015-500G
TCIAS0015-25GEA 132 DKK
TCIAS0015-25G TCIAS0015-500G
Methylsalicylat ≥99.0% (by GC)
Methylsalicylat
Formel: 2-(HO)C₆H₄CO₂CH₃
Molekylvægt: 152.15 g/mol
Kogepunkt: 224°C
Smeltepunkt: –9°C
Flammepunkt: 97°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002214
CAS-nummer: 119-36-8
EINECS: 204-317-7
Merck Index: 14,06120
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e9d68e443778019f895096cc9c89cc169c0b45d&vid=1e9d68e443778019f895096cc9c89cc169c0b45d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Acid to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR