Ethyl linoleat ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate
L0135-5ML
L0135-25ML
TCIAL0135-5MLEA
862
DKK
TCIAL0135-5ML
TCIAL0135-25ML
Ethyl linoleat ≥97.0% (by GC)
Ethyl linoleat
Formel:
C₂₀H₃₆O₂ Kogepunkt: 193°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00009535 CAS-nummer: 544-35-4 EINECS: 208-868-4 Merck Index: 14,03820 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...