Acetamid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0007-500G A0007-25G
TCIAA0007-500GEA 1900 DKK
TCIAA0007-500G TCIAA0007-25G
Acetamid ≥98.0% (by GC)
Acetamid
Formel: C₂H₅NO
Kogepunkt: 221,1 °C (1013 hPa)
Smeltepunkt: 78…81 °C
Massefylde: 1,14 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008023
CAS-nummer: 60-35-5
EINECS: 200-473-5
Merck Index: 14,00043
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42217f03134d8a5ed18aed0d37ee8372137faa4a&vid=42217f03134d8a5ed18aed0d37ee8372137faa4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR